Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AKAP1
- Entrez Gene ID
-
8165
- Gene Name
- A-kinase anchoring protein 1
- Gene Aliases
-
AKAP, AKAP121, AKAP149, AKAP84, D-AKAP1, PPP1R43, PRKA1, SAKAP84, TDRD17
- Chromosome Location
-
Chr.17: 57085192 - 57121349 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U34074.1 |
- |
2 - 3 |
1774 |
83 |
View Full Product Details