The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001131015.1 | NP_001124487.1 | 13 - 14 | 2066 | 88 |
NM_001131016.1 | NP_001124488.1 | 12 - 13 | 2194 | 88 | |
NM_001131017.1 | NP_001124489.1 | 13 - 14 | 2011 | 88 | |
NM_001131018.1 | NP_001124490.1 | 12 - 13 | 1954 | 88 | |
NM_001257976.1 | NP_001244905.1 | 11 - 12 | 2078 | 88 | |
NM_012127.2 | NP_036259.2 | 12 - 13 | 2234 | 88 | |
XM_005251888.3 | XP_005251945.2 | 12 - 13 | 2065 | 88 | |
XM_005251891.3 | XP_005251948.2 | 13 - 14 | 1897 | 88 | |
XM_005251892.3 | XP_005251949.2 | 13 - 14 | 1882 | 88 | |
XM_005251893.4 | XP_005251950.4 | 12 - 13 | 1820 | 88 | |
XM_006717039.3 | XP_006717102.2 | 11 - 12 | 1973 | 88 | |
XM_011518481.1 | XP_011516783.1 | 12 - 13 | 2050 | 88 | |
XM_011518482.2 | XP_011516784.2 | 11 - 12 | 1988 | 88 | |
XM_011518483.1 | XP_011516785.1 | 11 - 12 | 2105 | 88 | |
XM_017014594.1 | XP_016870083.1 | 12 - 13 | 2518 | 88 | |
XM_017014595.1 | XP_016870084.1 | 12 - 13 | 2162 | 88 | |
XM_017014596.1 | XP_016870085.1 | 13 - 14 | 2009 | 88 | |
GenBank mRNA | AB030835.1 | - | 12 - 13 | 2147 | 88 |
AF159025.1 | - | 11 - 12 | 2026 | 88 | |
AK292713.1 | - | 12 - 13 | 2185 | 88 | |
AK303288.1 | - | 13 - 14 | 1994 | 88 | |
AK303636.1 | - | 11 - 12 | 2061 | 88 | |
AK316393.1 | - | 10 - 11 | 1790 | 88 | |
BC004119.2 | - | 13 - 14 | 1851 | 88 | |
BC021163.2 | - | 12 - 13 | 1933 | 88 | |
EF441870.1 | - | 5 - 6 | 740 | 88 |