The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008395.3 | NP_001008396.1 | 1 - 2 | 79 | 102 |
NM_001318236.1 | NP_001305165.1 | 1 - 2 | 79 | 102 | |
NR_134526.1 | 1 - 2 | 79 | 102 | ||
NR_134527.1 | 1 - 2 | 79 | 102 | ||
XM_017012198.1 | XP_016867687.1 | 3 - 4 | 466 | 102 | |
XM_017012199.1 | XP_016867688.1 | 3 - 4 | 466 | 102 | |
GenBank mRNA | BC063401.1 | - | 1 - 2 | 102 | |
BC105609.1 | - | 1 - 2 | 102 | ||
BG715626.1 | - | 1 - 2 | 76 | 102 | |
BI598677.1 | - | 1 - 2 | 82 | 102 | |
CU450474.1 | - | 1 - 2 | 102 |