The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011520775.2 | XP_011519077.1 | 2 - 3 | 309 | 107 |
GenBank mRNA | BG680355.1 | - | 2 - 3 | 283 | 107 |