Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMTOR4
Entrez Gene ID
389541
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 4
Gene Aliases
C7orf59
Chromosome Location
Chr.7: 100130969 - 100154212 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008395.3 NP_001008396.1 3 - 4 275 67
NM_001318237.1 NP_001305166.1 2 - 3 194 67
NR_134526.1 3 - 4 261 67
NR_134527.1 4 - 5 317 67
NR_134528.1 3 - 4 274 67
NR_134529.1 3 - 4 254 67
XM_017012198.1 XP_016867687.1 5 - 6 662 67
GenBank mRNA AA772120.1 - 1 - 2 67
AJ714519.1 - 3 - 4 255 67
BC063401.1 - 3 - 4 255 67
BC105609.1 - 3 - 4 265 67
BC130553.1 - 3 - 4 222 67
BC130559.1 - 3 - 4 222 67
BE733059.1 - 2 - 3 185 67
BF978089.1 - - 274 67
BG715626.1 - 3 - 4 271 67
BG819624.1 - 3 - 4 235 67
BI598677.1 - 3 - 4 278 67
BI771662.1 - 2 - 3 180 67
CU450474.1 - 3 - 4 328 67
DB539334.2 - 2 - 3 197 67

View Full Product Details