Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IGF2R
Entrez Gene ID
3482
Gene Name
insulin like growth factor 2 receptor
Gene Aliases
CD222, CI-M6PR, CIMPR, M6P-R, M6P/IGF2R, MPR 300, MPR1, MPR300, MPRI
Chromosome Location
Chr.6: 159969099 - 160111504 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000876.3 NP_000867.2 33 - 34 4839 85
GenBank mRNA AB209668.1 - 32 - 33 4462 85
J03528.1 - 33 - 34 4838 85
Y00285.1 - 33 - 34 4838 85

View Full Product Details