Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX9
Entrez Gene ID
51429
Gene Name
sorting nexin 9
Gene Aliases
SDP1, SH3PX1, SH3PXD3A, WISP
Chromosome Location
Chr.6: 157716046 - 157945077 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016224.4 NP_057308.1 12 - 13 1554 70
XM_005267015.2 XP_005267072.1 14 - 15 1632 70
XM_011535886.2 XP_011534188.1 9 - 10 1093 70
GenBank mRNA AB209736.1 - 8 - 9 885 70
AF076957.1 - 7 - 8 774 70
AF121859.1 - 12 - 13 1314 70
AF131214.1 - 12 - 13 1333 70
AF172847.1 - 12 - 13 1434 70
AK026637.1 - 9 - 10 1099 70
AK127345.1 - 9 - 10 1409 70
AK314360.1 - 12 - 13 1388 70
BC001084.2 - 3 - 4 355 70
BC005022.1 - 12 - 13 1461 70

View Full Product Details