The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001972.3 | NP_001963.1 | 1 - 2 | 187 | 101 |
XM_011527775.1 | XP_011526077.1 | 2 - 3 | 864 | 101 | |
XM_011527776.1 | XP_011526078.1 | 2 - 3 | 203 | 101 | |
GenBank mRNA | BC074816.2 | - | 1 - 2 | 104 | 101 |
BC074817.2 | - | 1 - 2 | 104 | 101 | |
CD613590.1 | - | 2 - 3 | 208 | 101 | |
M34379.1 | - | 1 - 2 | 105 | 101 | |
X05875.1 | - | 1 - 2 | 105 | 101 |