Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPS8L2
Entrez Gene ID
64787
Gene Name
EPS8 like 2
Gene Aliases
EPS8R2
Chromosome Location
Chr.11: 705217 - 727727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022772.3 NP_073609.2 15 - 16 1700 100
XM_017018130.1 XP_016873619.1 16 - 17 1917 100
XM_017018131.1 XP_016873620.1 15 - 16 1788 100
XM_017018132.1 XP_016873621.1 16 - 17 1758 100
GenBank mRNA AF318331.1 - 3 - 4 869 100
AK025588.1 - 15 - 16 1618 100
AK025824.1 - 15 - 16 1618 100
AK027765.1 - 3 - 4 512 100
AK094539.1 - 7 - 8 796 100
AK122903.1 - 12 - 13 1896 100
AK222903.1 - 15 - 16 1613 100
AK225311.1 - 15 - 16 1618 100
AY074929.1 - 15 - 16 1725 100
BC080636.1 - 15 - 16 1704 100
BC093878.1 - 14 - 15 1493 100
BC101481.1 - 14 - 15 1493 100
BC143242.1 - 14 - 15 1541 100

View Full Product Details