The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261.3 | NP_001252.1 | 4 - 5 | 557 | 67 |
XM_017014184.1 | XP_016869673.1 | 4 - 5 | 617 | 67 | |
GenBank mRNA | AB451289.1 | - | 4 - 5 | 434 | 67 |
AB451421.1 | - | 4 - 5 | 434 | 67 | |
AK313828.1 | - | 4 - 5 | 486 | 67 | |
BC001968.1 | - | 4 - 5 | 494 | 67 | |
BT019903.1 | - | 4 - 5 | 434 | 67 | |
L25676.1 | - | 4 - 5 | 498 | 67 | |
X80230.1 | - | 4 - 5 | 532 | 67 |