The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014648.3 | NP_055463.1 | 32 - 33 | 4060 | 68 |
XM_005247914.3 | XP_005247971.1 | 32 - 33 | 3859 | 68 | |
XM_005247915.3 | XP_005247972.1 | 32 - 33 | 3829 | 68 | |
XM_005247916.3 | XP_005247973.1 | 33 - 34 | 4001 | 68 | |
XM_005247917.3 | XP_005247974.1 | 32 - 33 | 3790 | 68 | |
XM_005247918.3 | XP_005247975.1 | 32 - 33 | 4277 | 68 | |
XM_005247919.3 | XP_005247976.1 | 31 - 32 | 3411 | 68 | |
XM_017007540.1 | XP_016863029.1 | 32 - 33 | 3906 | 68 | |
GenBank mRNA | AF279370.1 | - | 17 - 18 | 1718 | 68 |
AK023138.1 | - | 32 - 33 | 3828 | 68 | |
AK294967.1 | - | 21 - 22 | 2123 | 68 | |
AY227652.1 | - | 19 - 20 | 1971 | 68 | |
AY227654.1 | - | 30 - 31 | 4496 | 68 | |
BC063882.1 | - | 32 - 33 | 3813 | 68 |