Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNDC3B
Entrez Gene ID
64778
Gene Name
fibronectin type III domain containing 3B
Gene Aliases
FAD104, PRO4979, YVTM2421
Chromosome Location
Chr.3: 172039628 - 172400703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135095.1 NP_001128567.1 20 - 21 2462 109
NM_022763.3 NP_073600.3 20 - 21 2535 109
XM_017007062.1 XP_016862551.1 19 - 20 2233 109
XM_017007063.1 XP_016862552.1 15 - 16 1719 109
GenBank mRNA AB098597.1 - 19 - 20 2420 109
AF543840.1 - 2 - 3 236 109
AK027052.1 - 4 - 5 518 109
AK075220.1 - 10 - 11 1157 109
AK127826.1 - 20 - 21 2515 109
AK223599.1 - 20 - 21 2566 109
AK314478.1 - 20 - 21 2462 109
AL157482.1 - 2 - 3 183 109
AY358146.1 - 13 - 14 1408 109
BC033635.2 - 8 - 9 1010 109
BC039297.1 - 20 - 21 2426 109
BX648340.1 - 20 - 21 2457 109

View Full Product Details