Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNDC3B
Entrez Gene ID
64778
Gene Name
fibronectin type III domain containing 3B
Gene Aliases
FAD104, PRO4979, YVTM2421
Chromosome Location
Chr.3: 172039628 - 172400703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135095.1 NP_001128567.1 24 - 25 3274 145
NM_022763.3 NP_073600.3 24 - 25 3347 145
XM_017007062.1 XP_016862551.1 23 - 24 3045 145
XM_017007063.1 XP_016862552.1 19 - 20 2531 145
GenBank mRNA AB098597.1 - 23 - 24 3232 145
AF543840.1 - 6 - 7 1048 145
AK027052.1 - 8 - 9 1330 145
AK075220.1 - 14 - 15 1969 145
AK127826.1 - 24 - 25 3327 145
AK223599.1 - 24 - 25 3378 145
AK314478.1 - 24 - 25 3274 145
AL157482.1 - 6 - 7 995 145
AY358146.1 - 17 - 18 2220 145
BC033635.2 - 12 - 13 1822 145
BC039297.1 - 24 - 25 3238 145
BX648340.1 - 24 - 25 3269 145

View Full Product Details