The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135095.1 | NP_001128567.1 | 15 - 16 | 1885 | 80 |
NM_022763.3 | NP_073600.3 | 15 - 16 | 1958 | 80 | |
XM_017007062.1 | XP_016862551.1 | 14 - 15 | 1656 | 80 | |
XM_017007063.1 | XP_016862552.1 | 10 - 11 | 1142 | 80 | |
XM_017007064.1 | XP_016862553.1 | 15 - 16 | 1938 | 80 | |
GenBank mRNA | AB098597.1 | - | 14 - 15 | 1843 | 80 |
AK075220.1 | - | 5 - 6 | 580 | 80 | |
AK127826.1 | - | 15 - 16 | 1938 | 80 | |
AK223599.1 | - | 15 - 16 | 1989 | 80 | |
AK314478.1 | - | 15 - 16 | 1885 | 80 | |
AY358146.1 | - | 8 - 9 | 831 | 80 | |
AY358367.1 | - | 15 - 16 | 1927 | 80 | |
BC033635.2 | - | 3 - 4 | 433 | 80 | |
BC039297.1 | - | 15 - 16 | 1849 | 80 | |
BX648340.1 | - | 15 - 16 | 1880 | 80 |