The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030767.5 | NP_110394.3 | 1 - 2 | 55 | 71 |
XM_005252244.2 | XP_005252301.1 | 3 - 4 | 227 | 71 | |
XM_005252245.1 | XP_005252302.1 | 2 - 3 | 85 | 71 | |
GenBank mRNA | BC055285.1 | - | 1 - 2 | 55 | 71 |