The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317950.1 | NP_001304879.1 | 2 - 3 | 492 | 64 |
NM_030767.5 | NP_110394.3 | 2 - 3 | 437 | 64 | |
XM_005252244.2 | XP_005252301.1 | 4 - 5 | 609 | 64 | |
XM_005252245.1 | XP_005252302.1 | 3 - 4 | 467 | 64 | |
XM_005252247.4 | XP_005252304.1 | 2 - 3 | 488 | 64 | |
XM_005252248.2 | XP_005252305.1 | 2 - 3 | 605 | 64 | |
XM_006717294.1 | XP_006717357.1 | 2 - 3 | 421 | 64 | |
XM_006717295.2 | XP_006717358.1 | 2 - 3 | 605 | 64 | |
XM_011519064.2 | XP_011517366.1 | 2 - 3 | 605 | 64 | |
XM_011519066.2 | XP_011517368.1 | 2 - 3 | 605 | 64 | |
GenBank mRNA | AB075848.1 | - | 2 - 3 | 610 | 64 |
AK126136.1 | - | 2 - 3 | 421 | 64 | |
AK131082.2 | - | 2 - 3 | 492 | 64 | |
AK160382.1 | - | 2 - 3 | 492 | 64 | |
AY703043.1 | - | 1 - 2 | 410 | 64 | |
BC055285.1 | - | 2 - 3 | 437 | 64 |