The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306079.1 | NP_001293008.1 | 41 - 42 | 6454 | 74 |
NM_004341.4 | NP_004332.2 | 42 - 43 | 6643 | 74 | |
XM_005264555.3 | XP_005264612.1 | 43 - 44 | 6827 | 74 | |
XM_006712101.2 | XP_006712164.1 | 42 - 43 | 6638 | 74 | |
GenBank mRNA | AK226105.1 | - | 41 - 42 | 6436 | 74 |
BC014178.2 | - | 17 - 18 | 2360 | 74 | |
BC065510.1 | - | 42 - 43 | 6643 | 74 | |
D78586.1 | - | 42 - 43 | 6507 | 74 |