The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033559.2 | NP_001028731.1 | 1 - 2 | 116 | 94 |
NM_001033560.1 | NP_001028732.1 | 1 - 2 | 116 | 94 | |
NM_130810.3 | NP_570722.2 | 1 - 2 | 116 | 94 | |
GenBank mRNA | AF337549.1 | - | 1 - 2 | 116 | 94 |