The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018968.3 | NP_061841.2 | 8 - 9 | 715 | 63 |
XM_011510366.2 | XP_011508668.1 | 6 - 7 | 752 | 63 | |
XM_017004361.1 | XP_016859850.1 | 8 - 9 | 677 | 63 | |
XM_017004362.1 | XP_016859851.1 | 8 - 9 | 683 | 63 | |
XM_017004363.1 | XP_016859852.1 | 8 - 9 | 864 | 63 | |
XM_017004364.1 | XP_016859853.1 | 5 - 6 | 4005 | 63 | |
XM_017004365.1 | XP_016859854.1 | 8 - 9 | 678 | 63 | |
XM_017004366.1 | XP_016859855.1 | 8 - 9 | 678 | 63 | |
XM_017004367.1 | XP_016859856.1 | 9 - 10 | 753 | 63 | |
XM_017004368.1 | XP_016859857.1 | 8 - 9 | 678 | 63 | |
XM_017004369.1 | XP_016859858.1 | 8 - 9 | 678 | 63 | |
XM_017004370.1 | XP_016859859.1 | 8 - 9 | 678 | 63 | |
XM_017004371.1 | XP_016859860.1 | 8 - 9 | 679 | 63 | |
XM_017004372.1 | XP_016859861.1 | 8 - 9 | 679 | 63 | |
XM_017004373.1 | XP_016859862.1 | 3 - 4 | 1319 | 63 | |
GenBank mRNA | AJ003029.1 | - | 8 - 9 | 717 | 63 |