The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145345.2 | NP_663320.2 | 3 - 4 | 437 | 72 |
GenBank mRNA | AF520989.1 | - | 2 - 3 | 146 | 72 |
AF521017.1 | - | 2 - 3 | 147 | 72 | |
AK097368.1 | - | 3 - 4 | 535 | 72 | |
AK291503.1 | - | 2 - 3 | 142 | 72 |