Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNHD1
Entrez Gene ID
144132
Gene Name
dynein heavy chain domain 1
Gene Aliases
C11orf47, CCDC35, DHCD1, DNHD1L
Chromosome Location
Chr.11: 6497296 - 6572025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144666.2 NP_653267.2 36 - 37 12916 73
GenBank mRNA AB290171.1 - 23 - 24 9616 73
AK057314.1 - 3 - 4 1435 73
AK128064.1 - 4 - 5 2558 73
AL137690.1 - 1 - 2 513 73
AL834222.1 - 3 - 4 2376 73
BC082757.1 - 5 - 6 1792 73
BC117301.1 - 3 - 4 1260 73
BC117303.1 - 3 - 4 1260 73
BC143910.1 - 3 - 4 1230 73

View Full Product Details