The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127207.1 | NP_001120679.1 | 13 - 14 | 2281 | 84 |
NM_014140.3 | NP_054859.2 | 13 - 14 | 2467 | 84 | |
XM_005246631.2 | XP_005246688.1 | 13 - 14 | 2405 | 84 | |
XM_005246632.1 | XP_005246689.1 | 13 - 14 | 2457 | 84 | |
XM_017004228.1 | XP_016859717.1 | 13 - 14 | 2270 | 84 | |
GenBank mRNA | AF082179.1 | - | 12 - 13 | 2237 | 84 |
AF432223.1 | - | 13 - 14 | 2277 | 84 | |
AK000117.1 | - | 9 - 10 | 1137 | 84 | |
AK023780.1 | - | 13 - 14 | 2407 | 84 | |
AL122076.1 | - | 11 - 12 | 1701 | 84 | |
BC016482.1 | - | 12 - 13 | 2228 | 84 | |
BC029994.1 | - | 10 - 11 | 1663 | 84 | |
BC043341.1 | - | 13 - 14 | 2254 | 84 |