The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184730.2 | NP_001171659.1 | 1 - 2 | 278 | 88 |
XM_005260430.2 | XP_005260487.1 | 1 - 2 | 256 | 88 | |
GenBank mRNA | AK302093.1 | - | 1 - 2 | 236 | 88 |