Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMARCAL1
Entrez Gene ID
50485
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1
Gene Aliases
HARP, HHARP
Chromosome Location
Chr.2: 216412414 - 216483053 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005246632.1 XP_005246689.1 1 - 2 220 66

View Full Product Details