Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIER2
Entrez Gene ID
54531
Gene Name
MIER family member 2
Gene Aliases
KIAA1193, Mi-er2
Chromosome Location
Chr.19: 305573 - 344793 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017550.1 NP_060020.1 8 - 9 815 71
XM_005259582.4 XP_005259639.1 8 - 9 1399 71
XM_006722769.3 XP_006722832.1 8 - 9 1403 71
XM_011528072.2 XP_011526374.1 8 - 9 1401 71
XM_011528073.1 XP_011526375.1 8 - 9 817 71
XM_011528074.2 XP_011526376.1 8 - 9 1405 71
XM_011528075.2 XP_011526377.1 8 - 9 1922 71
XM_011528077.2 XP_011526379.1 9 - 10 3763 71
XM_011528081.2 XP_011526383.1 6 - 7 659 71
XM_011528082.2 XP_011526384.1 6 - 7 590 71
GenBank mRNA AB033019.1 - - 803 71
BC028203.1 - 8 - 9 815 71

View Full Product Details