Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APPL1
Entrez Gene ID
26060
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1
Gene Aliases
APPL, DIP13alpha, MODY14
Chromosome Location
Chr.3: 57227737 - 57273471 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012096.2 NP_036228.1 5 - 6 516 98
XM_011533583.2 XP_011531885.1 6 - 7 569 98
GenBank mRNA AF169797.1 - 5 - 6 427 98
AF424738.1 - 5 - 6 425 98
AK299008.1 - 6 - 7 558 98
AK310716.1 - 5 - 6 464 98
BC028599.2 - 5 - 6 516 98

View Full Product Details