Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A2
Entrez Gene ID
6567
Gene Name
solute carrier family 16 member 2
Gene Aliases
AHDS, DXS128, DXS128E, MCT 7, MCT 8, MCT7, MCT8, MRX22, XPCT
Chromosome Location
Chr.X: 74421493 - 74533929 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006517.4 NP_006508.2 2 - 3 717 60
GenBank mRNA AB085789.1 - 2 - 3 659 60
U05315.1 - 2 - 3 749 60

View Full Product Details