The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320865.1 | NP_001307794.1 | 2 - 3 | 180 | 73 |
NM_198559.1 | NP_940961.1 | 2 - 3 | 147 | 73 | |
XM_011511148.2 | XP_011509450.1 | 2 - 3 | 279 | 73 | |
GenBank mRNA | BC052750.1 | - | 2 - 3 | 147 | 73 |