The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100176.1 | NP_001093646.1 | 1 - 2 | 216 | 66 |
NM_013312.2 | NP_037444.2 | 1 - 2 | 216 | 66 | |
XM_005259873.2 | XP_005259930.1 | 1 - 2 | 216 | 66 | |
XM_005259874.2 | XP_005259931.1 | 1 - 2 | 216 | 66 | |
XM_011527943.2 | XP_011526245.1 | 1 - 2 | 174 | 66 | |
XM_017026720.1 | XP_016882209.1 | 1 - 2 | 152 | 66 | |
GenBank mRNA | AF044924.1 | - | 1 - 2 | 119 | 66 |
BC012443.1 | - | 1 - 2 | 139 | 66 | |
DB474423.1 | - | 1 - 2 | 216 | 66 |