The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011530405.2 | XP_011528707.1 | 14 - 14 | 7629 | 128 |
XM_011530406.2 | XP_011528708.1 | 14 - 14 | 7095 | 128 | |
XM_011530407.2 | XP_011528709.1 | 14 - 14 | 7502 | 128 | |
XM_011530408.1 | XP_011528710.1 | 14 - 14 | 7093 | 128 | |
XM_011530410.2 | XP_011528712.1 | 12 - 12 | 7297 | 128 | |
XM_017028960.1 | XP_016884449.1 | 15 - 15 | 8027 | 128 | |
XM_017028961.1 | XP_016884450.1 | 14 - 14 | 7419 | 128 | |
XM_017028962.1 | XP_016884451.1 | 14 - 14 | 7354 | 128 | |
XM_017028963.1 | XP_016884452.1 | 14 - 14 | 7018 | 128 | |
XM_017028964.1 | XP_016884453.1 | 14 - 14 | 7090 | 128 | |
XM_017028965.1 | XP_016884454.1 | 8 - 8 | 6864 | 128 |