Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENTPD2
Entrez Gene ID
954
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
CD39L1, NTPDase-2
Chromosome Location
Chr.9: 137048098 - 137054051 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246.3 NP_001237.1 4 - 5 604 129
NM_203468.2 NP_982293.1 4 - 5 604 129
XM_011519212.2 XP_011517514.1 3 - 4 405 129
GenBank mRNA AF144748.1 - 4 - 5 550 129
BC035738.1 - 4 - 5 598 129
EF495152.1 - 4 - 5 550 129
U91510.1 - 4 - 5 571 129

View Full Product Details