Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENTPD2
Entrez Gene ID
954
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
CD39L1, NTPDase-2
Chromosome Location
Chr.9: 137048098 - 137054051 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246.3 NP_001237.1 5 - 6 834 116
NM_203468.2 NP_982293.1 5 - 6 834 116
XM_011519212.2 XP_011517514.1 4 - 5 635 116
GenBank mRNA AF144748.1 - 5 - 6 780 116
BC035738.1 - 5 - 6 828 116
EF495152.1 - 5 - 6 780 116
U91510.1 - 5 - 6 801 116

View Full Product Details