The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014228.4 | NP_055043.2 | 7 - 8 | 1420 | 67 |
XM_017009767.1 | XP_016865256.1 | 8 - 9 | 1327 | 67 | |
XM_017009768.1 | XP_016865257.1 | 8 - 9 | 1403 | 67 | |
XM_017009769.1 | XP_016865258.1 | 8 - 9 | 2934 | 67 | |
XM_017009770.1 | XP_016865259.1 | 8 - 9 | 1054 | 67 | |
GenBank mRNA | AK096607.1 | - | 7 - 8 | 1324 | 67 |
AK308895.1 | - | 7 - 8 | 1196 | 67 | |
AK312309.1 | - | 7 - 8 | 1200 | 67 | |
BC069631.1 | - | 7 - 8 | 953 | 67 | |
BC093785.1 | - | 7 - 8 | 1029 | 67 | |
BC113425.1 | - | 7 - 8 | 1029 | 67 |