The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004377.3 | NP_004368.1 | 1 - 2 | 84 | 60 |
NM_152246.2 | NP_689452.1 | 1 - 2 | 84 | 60 | |
GenBank mRNA | AK291316.1 | - | 1 - 2 | 48 | 60 |
BG426428.1 | - | - | 69 | 60 | |
CB961274.1 | - | 1 - 2 | 111 | 60 | |
Y08682.1 | - | 1 - 2 | 36 | 60 | |
Y08683.1 | - | 1 - 2 | 36 | 60 |