Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTNNB1
Entrez Gene ID
1499
Gene Name
catenin beta 1
Gene Aliases
CTNNB, MRD19, armadillo
Chromosome Location
Chr.3: 41199451 - 41240448 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098209.1 NP_001091679.1 1 - 2 223 93
NM_001098210.1 NP_001091680.1 1 - 2 223 93
NM_001904.3 NP_001895.1 1 - 2 223 93
XM_006712985.1 XP_006713048.1 1 - 2 197 93
GenBank mRNA AK296790.1 - 1 - 2 169 93
AK314776.1 - 1 - 2 169 93
BC058926.1 - 1 - 2 165 93
DA216720.1 - 1 - 2 223 93
X87838.1 - 1 - 2 169 93
Z19054.1 - 1 - 2 155 93

View Full Product Details