The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004704.4 | NP_004695.1 | 12 - 13 | 1261 | 59 |
GenBank mRNA | AA465355.1 | - | 3 - 4 | 58 | |
AJ001340.1 | - | 12 - 13 | 1221 | 59 | |
AK313696.1 | - | 12 - 13 | 1261 | 59 | |
BC001113.1 | - | 12 - 13 | 1244 | 59 | |
BC009879.2 | - | 12 - 13 | 1226 | 59 | |
BC010048.1 | - | 12 - 13 | 1244 | 59 | |
BC021032.1 | - | 12 - 13 | 1244 | 59 | |
BC023662.2 | - | 12 - 13 | 1192 | 59 |