The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004704.4 | NP_004695.1 | 7 - 8 | 717 | 76 |
GenBank mRNA | AJ001340.1 | - | 7 - 8 | 677 | 76 |
AK313696.1 | - | 7 - 8 | 717 | 76 | |
BC001113.1 | - | 7 - 8 | 700 | 76 | |
BC009879.2 | - | 7 - 8 | 682 | 76 | |
BC010048.1 | - | 7 - 8 | 700 | 76 | |
BC021032.1 | - | 7 - 8 | 700 | 76 | |
BC023662.2 | - | 7 - 8 | 648 | 76 |