Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
Chromosome Location
Chr.10: 91923769 - 92030998 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003972.2 NP_003963.1 20 - 21 2958 70
XM_011540326.1 XP_011538628.1 20 - 21 3057 70
XM_011540327.2 XP_011538629.1 17 - 18 2723 70
XM_011540328.2 XP_011538630.1 17 - 18 2551 70
XM_011540329.2 XP_011538631.1 20 - 21 3168 70
XM_017016877.1 XP_016872366.1 19 - 20 3020 70
XM_017016878.1 XP_016872367.1 19 - 20 2850 70
XM_017016879.1 XP_016872368.1 19 - 20 2974 70
XM_017016880.1 XP_016872369.1 19 - 20 2978 70
XM_017016881.1 XP_016872370.1 18 - 19 2827 70
GenBank mRNA AB208843.1 - 7 - 8 1247 70
AF038362.1 - 20 - 21 2977 70
AJ001017.2 - 20 - 21 2872 70
BC112201.1 - 20 - 21 2942 70

View Full Product Details