Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
Chromosome Location
Chr.10: 91923769 - 92030998 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003972.2 NP_003963.1 26 - 27 3910 107
XM_011540326.1 XP_011538628.1 26 - 27 4009 107
XM_011540327.2 XP_011538629.1 23 - 24 3675 107
XM_011540328.2 XP_011538630.1 23 - 24 3503 107
XM_017016877.1 XP_016872366.1 25 - 26 3972 107
XM_017016878.1 XP_016872367.1 25 - 26 3802 107
XM_017016879.1 XP_016872368.1 25 - 26 3926 107
XM_017016880.1 XP_016872369.1 25 - 26 3930 107
XM_017016881.1 XP_016872370.1 24 - 25 3779 107
GenBank mRNA AB208843.1 - 14 - 15 2304 107
AF038362.1 - 26 - 27 3929 107
AJ001017.2 - 26 - 27 3824 107
AK303554.1 - 4 - 5 587 107
BC029930.1 - 2 - 3 305 107
BC112201.1 - 26 - 27 3894 107

View Full Product Details