The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018222.4 | NP_060692.2 | 12 - 13 | 1210 | 93 |
XM_005253015.3 | XP_005253072.1 | 12 - 13 | 1199 | 93 | |
GenBank mRNA | AF237771.1 | - | 12 - 13 | 1133 | 93 |
AF325830.1 | - | 12 - 13 | 1041 | 93 | |
AK001655.1 | - | 12 - 13 | 1090 | 93 | |
AK098504.1 | - | 11 - 12 | 1134 | 93 | |
BC014535.1 | - | 12 - 13 | 1069 | 93 | |
BC016713.1 | - | 12 - 13 | 1127 | 93 |