Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MORN1
Entrez Gene ID
79906
Gene Name
MORN repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 2321253 - 2391751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024848.2 NP_079124.1 13 - 14 1509 148
XM_005244798.2 XP_005244855.1 12 - 13 1373 148
XM_011542169.1 XP_011540471.1 13 - 14 1506 148
XM_011542176.1 XP_011540478.1 11 - 12 1218 148
GenBank mRNA AK024003.1 - 13 - 14 1312 148

View Full Product Details