Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VAT1L
Entrez Gene ID
57687
Gene Name
vesicle amine transport 1-like
Gene Aliases
-
Chromosome Location
Chr.16: 77788564 - 77980107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020927.2 NP_065978.1 8 - 9 1286 65
GenBank mRNA AB046796.1 - 8 - 9 1256 65
AK290153.1 - 8 - 9 1264 65
AL834240.1 - 7 - 8 901 65
BC033537.1 - 8 - 9 1264 65

View Full Product Details