The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194946.1 | NP_001181875.1 | 30 - 31 | 4551 | 61 |
NM_001194947.1 | NP_001181876.1 | 29 - 30 | 4494 | 61 | |
NM_001291157.1 | NP_001278086.1 | 29 - 30 | 4583 | 61 | |
NM_004953.4 | NP_004944.3 | 22 - 23 | 4042 | 61 | |
NM_182917.4 | NP_886553.3 | 28 - 29 | 4678 | 61 | |
NM_198241.2 | NP_937884.1 | 29 - 30 | 4530 | 61 | |
NM_198242.2 | NP_937885.1 | 25 - 26 | 4115 | 61 | |
NM_198244.2 | NP_937887.1 | 26 - 27 | 4292 | 61 | |
GenBank mRNA | AB210013.1 | - | 28 - 29 | 4669 | 61 |
AF104913.1 | - | 26 - 27 | 4154 | 61 | |
AF281070.1 | - | 28 - 29 | 4536 | 61 | |
AK131407.1 | - | 9 - 10 | 1229 | 61 | |
AK226160.1 | - | 29 - 30 | 4464 | 61 | |
AK294567.1 | - | 27 - 28 | 4161 | 61 | |
AK299763.1 | - | 26 - 27 | 4220 | 61 | |
AL832754.1 | - | 25 - 26 | 4004 | 61 | |
AY082886.1 | - | 28 - 29 | 4536 | 61 | |
BC007788.2 | - | 11 - 12 | 1398 | 61 | |
BC082764.1 | - | 23 - 24 | 3653 | 61 | |
BC140892.1 | - | 30 - 31 | 4393 | 61 | |
BC140896.1 | - | 29 - 30 | 4336 | 61 | |
BT007282.1 | - | 11 - 12 | 1397 | 61 | |
BX647812.1 | - | 29 - 30 | 4443 | 61 | |
D12686.1 | - | 22 - 23 | 4033 | 61 |