Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF4G1
Entrez Gene ID
1981
Gene Name
eukaryotic translation initiation factor 4 gamma 1
Gene Aliases
EIF-4G1, EIF4F, EIF4G, EIF4GI, P220, PARK18
Chromosome Location
Chr.3: 184314495 - 184335358 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194946.1 NP_001181875.1 9 - 10 925 114
NM_001194947.1 NP_001181876.1 8 - 9 868 114
NM_001291157.1 NP_001278086.1 8 - 9 957 114
NM_004953.4 NP_004944.3 1 - 2 413 114
NM_182917.4 NP_886553.3 7 - 8 1049 114
NM_198241.2 NP_937884.1 8 - 9 904 114
NM_198242.2 NP_937885.1 4 - 5 489 114
NM_198244.2 NP_937887.1 5 - 6 666 114
GenBank mRNA AB210013.1 - 7 - 8 1043 114
AF104913.1 - 5 - 6 525 114
AF281070.1 - 7 - 8 907 114
AK096719.1 - 6 - 7 554 114
AK128378.1 - 7 - 8 653 114
AK226160.1 - 8 - 9 838 114
AK294567.1 - 6 - 7 535 114
AK299763.1 - 5 - 6 594 114
AK308319.1 - 3 - 4 254 114
AL832754.1 - 4 - 5 378 114
AY082886.1 - 7 - 8 907 114
BC140892.1 - 9 - 10 767 114
BC140896.1 - 8 - 9 710 114
BX647812.1 - 8 - 9 817 114
DB088474.1 - 1 - 2 251 114

View Full Product Details