The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017415.1 | NP_001017415.1 | 3 - 4 | 615 | 139 |
NM_001017416.1 | NP_001017416.1 | 3 - 4 | 520 | 139 | |
NM_003368.4 | NP_003359.3 | 3 - 4 | 1045 | 139 | |
GenBank mRNA | AB014458.1 | - | 3 - 4 | 396 | 139 |
AB208893.1 | - | 3 - 4 | 677 | 139 | |
AF117386.1 | - | 3 - 4 | 532 | 139 | |
AK312423.1 | - | 3 - 4 | 708 | 139 | |
AL117575.1 | - | 2 - 3 | 367 | 139 | |
BC018745.1 | - | 3 - 4 | 1045 | 139 | |
BC032364.1 | - | 3 - 4 | 695 | 139 | |
BC050525.1 | - | 3 - 4 | 749 | 139 |