Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP1
Entrez Gene ID
7398
Gene Name
ubiquitin specific peptidase 1
Gene Aliases
UBP
Chromosome Location
Chr.1: 62436304 - 62451804 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017415.1 NP_001017415.1 1 - 2 252 93
GenBank mRNA BI463072.1 - 1 - 2 261 93

View Full Product Details