Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARS
Entrez Gene ID
833
Gene Name
cysteinyl-tRNA synthetase
Gene Aliases
CARS1, CYSRS, MGC:11246
Chromosome Location
Chr.11: 3000922 - 3057451 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014437.2 NP_001014437.1 11 - 12 1354 80
NM_001194997.1 NP_001181926.1 11 - 12 1354 80
NM_001751.5 NP_001742.1 10 - 11 1105 80
NM_139273.3 NP_644802.1 10 - 11 1105 80
NR_036542.1 11 - 12 1401 80
XM_006718341.3 XP_006718404.1 10 - 11 1128 80
XM_017018389.1 XP_016873878.1 10 - 11 1128 80
XM_017018390.1 XP_016873879.1 11 - 12 1368 80
XM_017018391.1 XP_016873880.1 8 - 9 1352 80
XM_017018392.1 XP_016873881.1 10 - 11 1290 80
GenBank mRNA AB208814.1 - 9 - 10 3279 80
AF288206.1 - 10 - 11 1038 80
AF288207.1 - 10 - 11 1038 80
AK096313.1 - 11 - 12 1368 80
AK222614.1 - 10 - 11 1046 80
AK289432.1 - 10 - 11 1078 80
AK295371.1 - 10 - 11 996 80
AK296763.1 - 11 - 12 1219 80
AK298514.1 - 11 - 12 1321 80
AK302644.1 - 11 - 12 1321 80
AK308005.1 - 11 - 12 1274 80
BC002880.2 - 10 - 11 1091 80
BT009913.1 - 10 - 11 1021 80
BX647906.1 - 11 - 12 1328 80
L06845.1 - 8 - 9 892 80

View Full Product Details