The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251902.1 | NP_001238831.1 | 4 - 5 | 662 | 93 |
NM_003985.4 | NP_003976.2 | 4 - 5 | 662 | 93 | |
XM_011524045.2 | XP_011522347.1 | 4 - 5 | 710 | 93 | |
XM_011524046.2 | XP_011522348.1 | 4 - 5 | 799 | 93 | |
XM_011524047.2 | XP_011522349.1 | 4 - 5 | 799 | 93 | |
XM_011524048.1 | XP_011522350.1 | 4 - 5 | 567 | 93 | |
GenBank mRNA | AK225403.1 | - | 4 - 5 | 545 | 93 |
AK310684.1 | - | 4 - 5 | 545 | 93 | |
BC035782.1 | - | 3 - 4 | 527 | 93 | |
U43408.1 | - | 4 - 5 | 546 | 93 |