Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FASN
Entrez Gene ID
2194
Gene Name
fatty acid synthase
Gene Aliases
FAS, OA-519, SDR27X1
Chromosome Location
Chr.17: 82078338 - 82098303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004104.4 NP_004095.4 11 - 12 1987 81
XM_011523538.2 XP_011521840.1 11 - 12 2055 81
GenBank mRNA AB209988.1 - 11 - 12 1982 81
AY451392.1 - 10 - 11 1870 81
BC063242.1 - 11 - 12 1987 81
U29344.1 - 11 - 12 1993 81

View Full Product Details