Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FASN
Entrez Gene ID
2194
Gene Name
fatty acid synthase
Gene Aliases
FAS, OA-519, SDR27X1
Chromosome Location
Chr.17: 82078338 - 82098303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004104.4 NP_004095.4 20 - 21 3338 61
XM_011523538.2 XP_011521840.1 20 - 21 3406 61
GenBank mRNA AB209988.1 - 20 - 21 3333 61
AY451392.1 - 19 - 20 3221 61
BC063242.1 - 20 - 21 3338 61
U26644.1 - 19 - 20 3215 61
U29344.1 - 20 - 21 3344 61

View Full Product Details